Sodium-DL-3-hydroxybutyrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt

A11613-06 A11613-14
AAA11613-06EA 38.99 USD
AAA11613-06 AAA11613-14
Sodium-DL-3-hydroxybutyrate 98%
Sodium-DL-3-hydroxybutyrate

Purity: 98% CAS No.: 150-83-4 Molecular Formula: C4H7NaO3 Formula Weight: 126.09 Melting Point: 161-167° Beilstein Reference: 4824866 MDL No.: MFCD00016716

Formula: C₄H₈O₃
Melting Pt: 161…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00016716
CAS Number: 150-83-4
EINECS: 205-774-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR