Molecular sieve A4 (0.4 nm, 4 Å), powder

Supplier: Thermo Scientific Chemicals

A11535-30 A11535-0I A11535-0B
AAA11535-30EA 65.49 USD
AAA11535-30 AAA11535-0I AAA11535-0B
Molecular sieve A4 (0.4 nm, 4 Å), powder
Molecular sieve A4 (0.4 nm, 4 Å)

MDL: MFCD00131613


CAS No.: 70955-01-0 MDL No.: MFCD00131613

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR