3-Bromo-p-toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11496-14 A11496-06 A11496-22
AAA11496-14EA 145 USD
AAA11496-14 AAA11496-06 AAAA11496-22
3-Bromo-p-toluic acid 98+%
3-Bromo-p-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 206…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00002488
CAS Number: 7697-26-9
EINECS: 231-712-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR