1-Chlorobutane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00001009
Beilstein Registry No.: 1730909
Purity: 99+%
CAS No.: 109-69-3
Molecular Formula: CH3(CH2)3Cl
Formula Weight: 92.57
Flash Point: -12°(10°F)
Physical Form: Liquid
Density: 0.886
Refractive Index: 1.4024
Odor: Pungent
Melting Point: -123°
Boiling Point: 77-78°
Beilstein Reference: 1730909
Merck Reference: 14,1560
UN No.: 1127
MDL No.: MFCD00001009
Formula:
CH₃(CH₂)₃Cl Boiling Pt: 77…78 °C Melting Pt: –123 °C Density: 0.886 Flash Pt: –12 °C (10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001009 CAS Number: 109-69-3 EINECS: 203-696-6 UN: 1127 ADR: 3,II Merck Index: 14,1560 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...