Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
A11450-22
A11450-36
A11450-0E
AAA11450-22EA
72.22
USD
AAA11450-22
AAA11450-36
AAA11450-0E
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 98%
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Fieser: 5,554
Purity: 98%
CAS No.: 140-89-6
Molecular Formula: C3H5KOS2
Formula Weight: 160.30
Flash Point: 205°(401°F)
Physical Form: Powder
Density: 1.558
Appearance: Yellow
Odor: Unpleasant
pH: 9-10
Melting Point: ca 205° dec.
Beilstein Reference: 3596974
Merck Reference: 14,7696
MDL No.: MFCD00004931
Formula:
C₃H₅KOS₂ Melting Pt: ca 205 °C (dec.) Density: 1.558 Flash Pt: 205 °C(401 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004931 CAS Number: 140-89-6 EINECS: 205-439-3 Merck Index: 14,7696 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...