D-(-)-Serine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-(-)-Serine, H-D-Ser-OH
A11353-22
A11353-06
A11353-14
AAA11353-22EA
427
USD
AAA11353-22
AAA11353-06
AAA11353-14
D-(-)-Serine 99%
D(-)-Serine
MDL: MFCD00004269
Beilstein Registry No.: 1721403
Fieser: 14,282
Optical Rotation: -13°C (c=5 in 5N HCl)
Purity: 99%
CAS No.: 312-84-5
Molecular Formula: C3H7NO3
Formula Weight: 105.09
Melting Point: 218-220° dec.
Beilstein Reference: 1721403
Merck Reference: 14,8460
Optical Rotation: -13° (c=5 in 5N HCl)
MDL No.: MFCD00004269
Formula:
C₃H₇NO₃ Melting Pt: 218…220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004269 CAS Number: 312-84-5 EINECS: 206-229-4 Merck Index: 14,8460 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...