(3-Aminopropyl)trimethoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane
A11284-0E
A11284-22
A11284-36
AAA11284-0EEA
763
USD
AAA11284-0E
AAA11284-22
AAA11284-36
(3-Aminopropyl)trimethoxysilane 97%
(3-Aminopropyl)trimethoxysilane
MDL: MFCD00008206
Beilstein Registry No.: 2038118
Purity: 97%
CAS No.: 13822-56-5
Molecular Formula: C6H17NO3Si
Formula Weight: 179.29
Flash Point: 90°(194°F)
Physical Form: Liquid
Density: 1.02
Refractive Index: 1.4240
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Odor: Amine-like
Boiling Point: 194°
Beilstein Reference: 2038118
UN No.: 2735
MDL No.: MFCD00008206
Formula:
H₂NCH₂CH₂CH₂Si(OCH₃)₃ MW: 179.29 g/mol Boiling Pt: 194 Density: 1.02 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008206 CAS Number: 13822-56-5 EINECS: 237-511-5 UN: 3267 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...