6-Chloropurine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine

A11202-06 A11202-14 A11202-03
AAA11202-06EA 80.6 USD
AAA11202-06 AAA11202-14 AAA11202-03
6-Chloropurine 99%
6-Chloropurine

MDL: MFCD00075825 Beilstein Registry No.: 5774


Purity: 99% CAS No.: 87-42-3 Molecular Formula: C5H3ClN4 Formula Weight: 154.56 Melting Point: 300° dec. Beilstein Reference: 5774 Merck Reference: 14,2161 MDL No.: MFCD00075825

Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Boiling Pt: 185…192 °C (2 mmHg)
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 14,2161

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR