S-Methylisothiouronium sulfate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulfate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate, S-Methylisothiourea sulfate, 2-Methyl-2-thiopseudourea sulfate, 2-Methyl-2-thiopseudourea hemisulfate

A11044-0E A11044-36 A11044-30 A11044-22
AAA11044-0EEA 353 USD
AAA11044-0E AAA11044-36 AAAA11044-30 AAA11044-22
S-Methylisothiouronium sulfate 98+%
S-Methylisothiouronium sulphate

Purity: 98+% CAS No.: 867-44-7 Molecular Formula: C4H12N4S2.H2SO4 Formula Weight: 278.38 Melting Point: ca 235° dec. Beilstein Reference: 3917217

Formula: C₄H₁₂N₄S₂·H₂SO₄
MW: 278.38 g/mol
Melting Pt: ∼ 235 °C (dec.)
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00013055
CAS Number: 867-44-7
EINECS: 212-759-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR