Fumaric acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00002700
Beilstein Registry No.: 605763
Fieser: 5,319
Purity: 99%
CAS No.: 110-17-8
Molecular Formula: HO2CCH=CHCO2H
Formula Weight: 116.07
Flash Point: 273°(523°F)
Odor: Odorless
pH: 2.1
Melting Point: ca 299° subl.
Beilstein Reference: 605763
Merck Reference: 14,4287
MDL No.: MFCD00002700
Formula:
C₄H₄O₄ Boiling Pt: 356 °C (1013 hPa) Melting Pt: ∼ 299 °C (subl.) Density: 1.625 Flash Pt: 273 °C (523 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002700 CAS Number: 110-17-8 EINECS: 203-743-0 Merck Index: 14,4287 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...