Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
A10882-22
A10882-06
A10882-14
AAA10882-22EA
1091
USD
AAA10882-22
AAA10882-06
AAA10882-14
Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Purity: 99%
CAS No.: 6994-25-8
Molecular Formula: C6H9N3O2
Formula Weight: 155.16
Melting Point: 104-107°
Beilstein Reference: 4964
Formula:
C₆H₉N₃O₂ Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005238 CAS Number: 6994-25-8 EINECS: 230-262-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...