EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Crystalline.
Purity: 98+%
CAS No.: 25952-53-8
Molecular Formula: C8H17N3.HCl
Formula Weight: 191.70
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: ca 113°
Beilstein Reference: 5764110
MDL No.: MFCD00012503
Formula:
C₈H₁₈ClN₃ Melting Pt: ∼ 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012503 CAS Number: 25952-53-8 EINECS: 247-361-2 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Melting Point | 112–114°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...