Tributyltin chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorotributylstannane, Tri-n-butyltin chloride, TBTC
A10746-30
A10746-18
A10746-0B
AAAA10746-30EA
218
USD
AAAA10746-30
AAAA10746-18
AAA10746-0B
Tributyltin chloride 96%
Tributyltin chloride
Purity: 96%
CAS No.: 1461-22-9
Molecular Formula: C12H27ClSn
Formula Weight: 325.51
Flash Point: 120°(248°F)
Refractive Index: 1.4910
Melting Point: -19°
Boiling Point: 171-173°/25mm
Beilstein Reference: 3535715
UN No.: 2788
MDL No.: MFCD00000521
Formula:
C₁₂H₂₇ClSn MW: 325.51 g/mol Boiling Pt: 171…173 °C (25 mmHg) Melting Pt: -9 Density: 1.200 Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000521 CAS Number: 1461-22-9 EINECS: 215-958-7 UN: 2788 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...