Lead(II) citrate trihydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead citrate, trihydrate

A10701-22
AAA10701-36EA 478.41 USD
AAA10701-36 AAA10701-22
Lead(II) citrate trihydrate 99%
Lead(II) citrate trihydrate

Soluble in cold water


Purity: 99% CAS No.: 6107-83-1 Molecular Formula: C12H10O14Pb3.3H2O Formula Weight: 1053.82 (999.77anhy) Solubility: Soluble in cold water UN No.: 2291 MDL No.: MFCD00150456

Formula: C₁₂H₁₆O₁₇Pb₃
Storage Temperature: Ambient
MDL Number: MFCD00150456
CAS Number: 6107-83-1
EINECS: 208-141-1
UN: 2291
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR