Ammonium formate, (max. 3% H₂O) 97% (by acidometric titration)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Formic acid ammonium salt
A10699-0I
A10699-0B
A10699-30
AAA10699-0IEA
410
USD
AAA10699-0I
AAA10699-0B
AAA10699-30
Ammonium formate, (max. 3% H₂O) 97% (by acidometric titration)
Ammonium formate
MDL: MFCD00013103
Beilstein Registry No.: 3625095
Fieser: 1,38 15,13 18,23 20,16
Purity: 97%
CAS No.: 540-69-2
Molecular Formula: CH5NO2
Formula Weight: 63.06
Odor: Ammonia-like
Melting Point: ca 116°
Beilstein Reference: 3625095
Merck Reference: 14,523
MDL No.: MFCD00013103
Formula:
HCO₂NH₄ MW: 63.06 g/mol Melting Pt: ∼ 116 °C Density: 1.28 Storage Temperature: Ambient |
MDL Number:
MFCD00013103 CAS Number: 540-69-2 EINECS: 208-753-9 Merck Index: 14,523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...