Pentaerythritol 98+%
Supplier: Thermo Scientific Chemicals
A10520-0B
A10520-0I
A10520-30
AAA10520-0BEA
43.5
USD
AAA10520-0B
AAA10520-0I
AAA10520-30
Pentaerythritol 98+%
Pentaerythritol
MDL: MFCD00004692
Beilstein Registry No.: 1679274
Purity: 98+%
CAS No.: 115-77-5
Molecular Formula: C5H12O4
Formula Weight: 136.15
Flash Point: 240°(464°F)
Melting Point: 260-264°
Boiling Point: 276°/30mm
Beilstein Reference: 1679274
Merck Reference: 14,7111
MDL No.: MFCD00004692
Formula:
C₅H₁₂O₄ MW: 136.15 g/mol Boiling Pt: 380 Melting Pt: 260…264 °C Density: 1.396 Flash Pt: 240 °C (464 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004692 CAS Number: 115-77-5 EINECS: 204-104-9 Merck Index: 14,7111 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...