3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

A10482-06 A10482-03
AAA10482-06EA 146 USD
AAA10482-06 AAA10482-03
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%
3-Hydroxy-4-methoxycinnamic acid

Purity: 98+% CAS No.: 537-73-5 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: ca 231° dec. Beilstein Reference: 2212760

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Melting Pt: ∼ 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR