Cyanoguanidine 99%
Supplier: Thermo Scientific Chemicals
MDL: MFCD00008066
Beilstein Registry No.: 605637
Fieser: 1,229
Purity: 99%
CAS No.: 461-58-5
Molecular Formula: C2H4N4
Formula Weight: 84.08
Physical Form: Powder
Density: 1.400
Appearance: White
Odor: Odorless
Melting Point: 208-211°
Beilstein Reference: 605637
Merck Reference: 14,3092
MDL No.: MFCD00008066
Formula:
(H₂N)₂CNCN Boiling Pt: 143 °C (1013 hPa) Melting Pt: 208…211 °C Density: 1.400 Storage Temperature: Ambient |
MDL Number:
MFCD00008066 CAS Number: 461-58-5 EINECS: 207-312-8 Merck Index: 14,3092 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...