Ninhydrin 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

A10409-09 A10409-14 A10409-18 A10409-30
AAA10409-09EA 40.33 USD
AAA10409-09 AAAA10409-14 AAA10409-18 AAA10409-30
Ninhydrin 99%
Ninhydrin

MDL: MFCD00003791 Beilstein Registry No.: 1910963 Fieser: 1,732 4,356

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 251 °C (dec.)
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 14,6554
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR