1,4-Dioxaspiro[4.5]decan-8-one 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

A10408-14 A10408-06 A10408-22
AAA10408-14EA 131 USD
AAA10408-14 AAA10408-06 AAA10408-22
1,4-Dioxaspiro[4.5]decan-8-one 97%
1,4-Dioxaspiro[4.5]decan-8-one

Purity: 97% CAS No.: 4746-97-8 Molecular Formula: C8H12O3 Formula Weight: 156.18 Melting Point: 72-75° Beilstein Reference: 117631

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR