D-(-)-Arabinose 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
A10357-30
A10357-09
A10357-18
AAA10357-30EA
360
USD
AAA10357-30
AAAA10357-09
AAA10357-18
D-(-)-Arabinose 99%
D(-)-Arabinose
MDL: MFCD00135608
Beilstein Registry No.: 1723079
Optical Rotation: -104°C (c=10 in water, 24h)
Purity: 99%
CAS No.: 10323-20-3
Molecular Formula: C5H10O5
Formula Weight: 150.13
Melting Point: 154-158°
Beilstein Reference: 1723079
Formula:
C₅H₁₀O₅ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135608 CAS Number: 10323-20-3 EINECS: 233-708-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...