Benzaldehyde 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10348-0I
AAA10348-0IEA
194
USD
AAA10348-0I
Benzaldehyde 99+%
Benzaldehyde
Fieser: 1,46 6,26
Purity: 99+%
CAS No.: 100-52-7
Molecular Formula: C7H6O
Formula Weight: 106.12
Flash Point: 62°(143°F)
Physical Form: Liquid
Density: 1.047
Refractive Index: 1.5450
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Almond-like
Melting Point: -26°
Boiling Point: 178-179°
Beilstein Reference: 471223
Merck Reference: 14,1058
UN No.: 1990
MDL No.: MFCD00003299
Formula:
C₆H₅CHO MW: 106.12 g/mol Boiling Pt: 178…179 °C Melting Pt: –26 °C Density: 1.047 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003299 CAS Number: 100-52-7 EINECS: 202-860-4 UN: 1990 ADR: "9",III Merck Index: 14,1058 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...