N,N-Dimethyl-1,4-phenylenediammonium dichloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A10343-22 A10343-36 A10343-14
AAA10343-22EA 264 USD
AAA10343-22 AAA10343-36 AAA10343-14
N,N-Dimethyl-1,4-phenylenediammonium dichloride 98%
N,N-Dimethyl-1,4-phenylenediammonium dichloride

MDL: MFCD00012991 Soluble in water


Purity: 98% CAS No.: 536-46-9 Molecular Formula: C8H12N2.2HCl Formula Weight: 209.12 Melting Point: 212° dec. Solubility: Soluble in water MDL No.: MFCD00012991

Formula: (CH₃)₂NC₆H₄NH₂·2HCl
Melting Pt: 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012991
CAS Number: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,03280

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR