n-Pentadecane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Pentadecane, Pentadecane
A10336-30
A10336-14
A10336-36
A10336-18
AAA10336-30EA
150.27
USD
AAA10336-30
AAAA10336-14
AAAA10336-36
AAA10336-18
n-Pentadecane 99%
n-Pentadecane
MDL: MFCD00008990
Beilstein Registry No.: 1698194
Purity: 99%
CAS No.: 629-62-9
Molecular Formula: CH3(CH2)13CH3
Formula Weight: 212.42
Flash Point: 132°(269°F)
Refractive Index: 1.4320
Melting Point: 9-10°
Boiling Point: 269-270°
Beilstein Reference: 1698194
MDL No.: MFCD00008990
Formula:
H₃C(CH₂)₁₃CH₃ Boiling Pt: 269…270 °C Melting Pt: 9…10 °C Density: 0.769 Flash Pt: 132 °C(269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008990 CAS Number: 629-62-9 EINECS: 211-098-1 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...