n-Hexadecane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexadecane, Cetane
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.
Purity: 99%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colourless
Melting Point: 18°
Boiling Point: 287°
Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water.
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142
Formula:
H₃C(CH₂)₁₄CH₃ Boiling Pt: 287 °C Melting Pt: 18 °C Density: 0.773 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008998 CAS Number: 544-76-3 EINECS: 208-878-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...