L-Methionine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
A10318-0E
A10318-36
AAA10318-0EEA
886
USD
AAA10318-0E
AAA10318-36
L-Methionine 98+%
L-Methionine
MDL: MFCD00063097
Beilstein Registry No.: 1722294
Optical Rotation: +23°C (c=2 in 2N HCl)
Purity: 98+%
CAS No.: 63-68-3
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Melting Point: ca 280° dec.
Beilstein Reference: 1722294
Merck Reference: 14,5975
Optical Rotation: +23° (c=2 in 2N HCl)
MDL No.: MFCD00063097
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Boiling Pt: 278 Melting Pt: 136 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 EINECS: 200-562-9 Merck Index: 14,5975 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...