3,3-Dimethylbutanone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone
A10289-36
A10289-22
AAA10289-36EA
374
USD
AAA10289-36
AAA10289-22
3,3-Dimethylbutanone 97%
3,3-Dimethylbutanone
MDL: MFCD00008846
Purity: 97%
CAS No.: 75-97-8
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 5°(41°F)
Refractive Index: 1.3970
Melting Point: -53°
Boiling Point: 105-106°
Merck Reference: 14,7440
UN No.: 1224
MDL No.: MFCD00008846
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 106 Melting Pt: -53 Density: 0.803 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008846 CAS Number: 75-97-8 EINECS: 200-920-4 UN: 1224 ADR: 3 III Merck Index: 14,7440 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...