Diisopropylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00008862
Beilstein Registry No.: 605284
Purity: 99+%
CAS No.: 108-18-9
Molecular Formula: [(CH3)2CH]2NH
Formula Weight: 101.19
Flash Point: -7°(19°F)
Physical Form: Liquid
Density: 0.718
Refractive Index: 1.3920
Odor: Ammonia-like
Melting Point: -61°
Boiling Point: 83-84°
Beilstein Reference: 605284
Merck Reference: 14,3196
UN No.: 1158
MDL No.: MFCD00008862
Formula:
C₆H₁₅N Boiling Pt: 83…84 °C Melting Pt: –61 °C Density: 0.718 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008862 CAS Number: 108-18-9 EINECS: 203-558-5 UN: 1158 ADR: 3,II Merck Index: 14,3196 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...