trans-Stilbene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene

A10232-36 A10232-22
AAA10232-36EA 658 USD
AAA10232-36 AAA10232-22
trans-Stilbene 98%
trans-Stilbene

MDL: MFCD00064300 Beilstein Registry No.: 1616740


Purity: 98% CAS No.: 103-30-0 Molecular Formula: C14H12 Formula Weight: 180.25 Odor: Odorless Melting Point: 122-125° Boiling Point: 305-307° Beilstein Reference: 1616740 Merck Reference: 14,8817 UN No.: 3077 MDL No.: MFCD00064300

Formula: C₁₄H₁₂
MW: 180.25 g/mol
Boiling Pt: 306
Melting Pt: 122…125 °C
Density: 1.14
Storage Temperature: Ambient
MDL Number: MFCD00064300
CAS Number: 103-30-0
EINECS: 203-098-5
UN: 3077
ADR: 9 III
Merck Index: 14,8817
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR