Tetraethylammonium tetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetrylammonium tetrafluoroborate
A10211-18
A10211-30
A10211-06
A10211-22
AAA10211-18EA
154.21
USD
AAA10211-18
AAA10211-30
AAAA10211-06
AAAA10211-22
Tetraethylammonium tetrafluoroborate 99%
Tetraethylammonium tetrafluoroborate
MDL: MFCD00011827
Beilstein Registry No.: 3917638
Purity: 99%
CAS No.: 429-06-1
Molecular Formula: C8H20BF4N
Formula Weight: 217.06
Melting Point: 370-380° dec.
Beilstein Reference: 3917638
UN No.: 2811
MDL No.: MFCD00011827
Formula:
C₈H₂₀BF₄N Melting Pt: 370…380 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011827 CAS Number: 429-06-1 EINECS: 207-055-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...