(S)-Methyl-2-aminopropanoate hydrochloride 99%
Supplier: Thermo Scientific Chemicals
A10178-06
A10178-14
A10178-22
AAA10178-06EA
31.2
USD
AAA10178-06
AAA10178-14
AAA10178-22
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride
Purity: 99%
CAS No.: 2491-20-5
Molecular Formula: C4H9NO2.HCl
Formula Weight: 139.58
Storage Temperature: 2°C to 8°C
Melting Point: 106-110°
Beilstein Reference: 3594033
Optical Rotation: +6.5° (c=10 in methanol)
MDL No.: MFCD00063663
Formula:
C4H10ClNO2 Melting Pt: 106…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063663 CAS Number: 2491-20-5 EINECS: 219-652-4 |
Specification Test Results
Optical rotation | +6.5° (c=10 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...