Trimethyl(methylcyclopentadienyl)platinum(IV)
Supplier: Thermo Scientific Chemicals
Synonyms:
Trimethyl (methylcyclopentadienyl) platinium (IV), Platinum, trimethyl(2-methyl-1,3-cyclopentadien-1-yl)-, TRIMETHYL(METHYLCYCLOPENTADIENYL)PLATINUM
098158-02
AA98158-02EA
417
USD
AA98158-02
Trimethyl(methylcyclopentadienyl)platinum(IV)
Trimethyl(methylcyclopentadienyl)platinum(IV)
CAS No.: 94442-22-5
Molecular Formula: C9H16Pt
Formula Weight: 319.32
Physical Form: Powder/Pieces
Sensitivity: Air Sensitive
Appearance: White to off-white
Melting Point: 28-31°
Boiling Point: 23°/0.053mm subl.
MDL No.: MFCD00079665
Formula:
C₉H₁₆Pt Boiling Pt: 23 °C (0,053 mmHg) (subl.) Melting Pt: 28…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079665 CAS Number: 94442-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...