Thulium, AAS Standard Solution, Specpure®, Tm 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
089889-AE
AA89889-AEEA
51.64
USD
AA89889-AE
AA89889-AP
Thulium, AAS Standard Solution, Specpure®, Tm 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Thulium Standards
Molecular Formula: Tm2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...