Potassium bromate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution

Supplier: Thermo Scientific Chemicals
Danger

088592-LQ
AA88592-LQEA 107 USD
AA88592-LQ AA88592-LS
Potassium bromate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Potassium bromate

MDL: MFCD00011359 Notes: One unit makes one liter of 0.1N solution after dilution


CAS No.: 7758-01-2 Molecular Formula: KBrO3 Formula Weight: 167.00 Physical Form: Liquid Appearance: Colorless UN No.: 3139 MDL No.: MFCD00011359

Formula: KBrO₃
MW: 167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 3139
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR