Gallium arsenide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
088458-09
088458-18
088458-04
AA88458-09EA
549
USD
AA88458-09
AA88458-18
AA88458-04
Gallium arsenide ≥99.999% (metals basis)
Gallium arsenide
Semiconductors (transistors, lasers, solar cells)
Purity: 99.999%
CAS No.: 1303-00-0
Molecular Formula: GaAs
Formula Weight: 144.64
Physical Form: 25mm & Down Polycrystalline Pieces
Density: 5.31
Melting Point: 1238°
Merck Reference: 14,4347
UN No.: 1557
MDL No.: MFCD00011017
Semiconductors (transistors, lasers, solar cells)
Formula:
AsGa Melting Pt: 1238 °C Density: 5.31 Storage Temperature: Ambient |
MDL Number:
MFCD00011017 CAS Number: 1303-00-0 EINECS: 215-114-8 UN: 1557 ADR: 6.1,II Merck Index: 14,4347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...