Barium titanate ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

088267-22 088267-36
AA88267-22EA 110 USD
AA88267-22 AA88267-36
Barium titanate ≥99.7% (metals basis)
Barium titanate

MDL: MFCD00003447


Purity: 99.7% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 2 Micron Powder Density: 5.85 Appearance: White Odor: Odorless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Formula: BaO₃Ti
Melting Pt: 1620 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR