Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Diameter 3-4mm (0.12-0.16in)

Supplier: Thermo Scientific Chemicals

088120-A1 088120-A7
AA88120-A1EA 144.36 USD
AA88120-A1 AA88120-A7 AA88120-30
Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Diameter 3-4mm (0.12-0.16in)
Storage Temperature: Ambient
MDL Number: MFCD00147627
CAS Number: 70955-01-0
UN: 3464
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR