Scandium, AAS standard solution, Specpure®, Sc 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
088093-AP
AA88093-APEA
168.4
USD
AA88093-AP
Scandium, AAS standard solution, Specpure®, Sc 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Scandium Standards
Sc₂O₃ in 5% HNO₃