Lanthanum, AAS Standard Solution, Specpure®, La 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

088074-AE
AA88074-AEEA 44.63 USD
AA88074-AE
Lanthanum, AAS Standard Solution, Specpure®, La 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Lanthanum Standards

Molecular Formula: La in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR