Europium, AAS standard solution, Specpure®, Eu 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
088064-AP
AA88064-APEA
130.93
USD
AA88064-AP
Europium, AAS standard solution, Specpure®, Eu 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Europium Standards
Molecular Formula: Eu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...