Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 1-2mm (0.04-0.08in)
Supplier: Thermo Scientific Chemicals
087956-A1
087956-A7
AA87956-A1EA
158.63
USD
AA87956-A1
AA87956-A7
AA87956-30
Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 1-2mm (0.04-0.08in)
CAS No.: 70955-01-0
Physical Form: Pellets
Sensitivity: Hygroscopic
Appearance: Pale brown
MDL No.: MFCD00131613
Storage Temperature:
Ambient |
MDL Number:
MFCD03457537 CAS Number: 70955-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...