Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III), Eu(TMHD)3
087898-06
087898-03
AA87898-06EA
224
USD
AA87898-06
AA87898-03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III)
CAS No.: 15522-71-1
Molecular Formula: C33H57EuO6
Formula Weight: 701.78
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 187-189°
MDL No.: MFCD00000019
Formula:
C₃₃H₅₇EuO₆ Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000019 CAS Number: 15522-71-1 EINECS: 239-565-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...