Tetraiodosilane ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
087303-06
087303-14
AA87303-06EA
156.99
USD
AA87303-06
AA87303-14
Tetraiodosilane ≥99% (metals basis)
Tetraiodosilane
MDL: MFCD00054041
Purity: 99%
CAS No.: 13465-84-4
Molecular Formula: I4Si
Formula Weight: 535.70
Physical Form: Crystalline, Packaged under argon
Density: 4.198
Appearance: White
Melting Point: 120°
Boiling Point: 290°
UN No.: 2923
MDL No.: MFCD00054041
Formula:
I₄Si Boiling Pt: 290 °C Melting Pt: 120 °C Density: 4,198 Storage Temperature: Ambient |
MDL Number:
MFCD00054041 CAS Number: 13465-84-4 EINECS: 236-706-2 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...