Potassium hexafluorozirconate 99%

Supplier: Thermo Scientific Chemicals
Danger

086102-36 086102-A3
AA86102-36EA 97.25 USD
AA86102-36 AA86102-A3
Potassium hexafluorozirconate 99%
Potassium hexafluorozirconate

For manufacture of metallic zirconium Slightly soluble in cold water, soluble in hot water


Purity: 99% CAS No.: 16923-95-8 Molecular Formula: K2ZrF6 Formula Weight: 283.41 Physical Form: -10 Mesh Density: 3.48 Solubility: Slightly soluble in cold water, soluble in hot water Merck Reference: 14,7639 UN No.: 3288 MDL No.: MFCD00042547


For manufacture of metallic zirconium

Formula: F₆K₂Zr
MW: 283.41 g/mol
Density: 3.48
Storage Temperature: Ambient
MDL Number: MFCD00042547
CAS Number: 16923-95-8
EINECS: 240-985-6
UN: 3288
ADR: 6.1 III
Merck Index: 14,7639

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR