Tin(IV) chloride ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannic chloride, Tin tetrachloride

071203-22 071203-14
AA71203-22EA 379 USD
AA71203-22 AA71203-14
Tin(IV) chloride ≥99.99% (metals basis)
Tin (IV) chloride

MDL: MFCD00011242 Fieser: 1,1111 14,304 15,311 17,335 18,354 19,338 20,373 21,422


Purity: 99.99% CAS No.: 7646-78-8 Molecular Formula: Cl4Sn Formula Weight: 260.50 Physical Form: Liquid Density: 2.26 Refractive Index: 1.512 Sensitivity: Moisture Sensitive Appearance: Colorless Melting Point: -33° Boiling Point: 114° Merck Reference: 14,8774 UN No.: 1827 MDL No.: MFCD00011242

Formula: Cl₄Sn
Boiling Pt: 114 °C
Melting Pt: –33 °C
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 14,8774

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR