Tetrallyltin ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetrallylstannane, Tetraallyltin
071158-04
071158-09
AA71158-04EA
74.8
USD
AA71158-04
AA71158-09
Tetrallyltin ≥96%
Tetrallyltin
MDL: MFCD00008637
Purity: 96%
CAS No.: 7393-43-3
Molecular Formula: Sn(CH2CH=CH2)4
Formula Weight: 282.98
Flash Point: 75°(167°F)
Physical Form: Liquid
Density: 1.179
Refractive Index: 1.5385
Sensitivity: Air Sensitive
Boiling Point: 69-70°/1.5mm
UN No.: 2788
MDL No.: MFCD00008637
Formula:
C₁₂H₂₀Sn Boiling Pt: 69…70 °C (1,5 mmHg) Density: 1,179 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008637 CAS Number: 7393-43-3 EINECS: 230-987-5 UN: 2788 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...