Diphenyltin dichloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dichlorodiphenylstannane
071139-04
071139-09
071139-18
AA71139-04EA
19.1
USD
AA71139-04
AA71139-09
AA71139-18
Diphenyltin dichloride ≥95%
Diphenyltin dichloride
MDL: MFCD00000516
Purity: 95%
CAS No.: 1135-99-5
Molecular Formula: C12H10Cl2Sn
Formula Weight: 343.81
Flash Point:
Formula:
C₁₂H₁₀Cl₂Sn Boiling Pt: 333…337 °C (dec.) Melting Pt: 41…43 °C Flash Pt: >150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000516 CAS Number: 1135-99-5 EINECS: 214-496-3 UN: 3146 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...