Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate

067104-03 067104-06
AA67104-03EA 128 USD
AA67104-03 AA67104-06
Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®
Scandium (III) nitrate

MDL: MFCD00011220 Freely soluble in water and alcohol.


Purity: 99.9% CAS No.: 13465-60-6 Molecular Formula: Sc(NO3)3.xH2O Formula Weight: 230.97(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Solubility: Freely soluble in water and alcohol Merck Reference: 14,8392 UN No.: 1477 MDL No.: MFCD00011220

Formula: Sc(NO₃)₃·xH₂O
MW: 230.97
Storage Temperature: Ambient
MDL Number: MFCD00011220
CAS Number: 13465-60-6
EINECS: 236-701-5
UN: 1477
ADR: 5.1 III
Merck Index: 14,8392

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR