Nickel(II) chloride hexahydrate ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

053131-30 053131-A1
AA53131-18EA 62.53 USD
AA53131-18 AA53131-30 AA53131-A1
Nickel(II) chloride hexahydrate ≥99.95% (metals basis)
Nickel (II) chloride hexahydrate

MDL: MFCD00149809 Fieser: 12,336 15,217 21,281


Purity: 99.95% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Physical Form: Crystalline Density: 1.92 Appearance: Green Odor: Odorless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809

Formula: NiCl₂·6H₂O
Melting Pt: 140 °C (-H₂O)
Density: 1.92
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR