Imidazole ≥99%, crystalline powder ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

047274-0E 047274-22 047274-36
AA47274-0EEA 495.47 USD
AA47274-0E AA47274-22 AA47274-36
Imidazole ≥99%, crystalline powder ACS
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 88…92 °C
Density: 1.03
Flash Pt: 145 °C (293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 III
Merck Index: 14,4912
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR